siRNA Details
Gene Symbol: PHLPP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289003.1 | NP_001275932.1 | 16, 17 | 3111 | ||
NM_015020.3 | NP_055835.2 | 17, 18 | 2965 | |||
GenBank mRNA | AB023148.2 | - | 16, 17 | 3111 | ||
AK001854.1 | - | 5, 6 | 674 | |||
BC035267.1 | - | 12, 13 | 1800 | |||
BC129927.1 | - | 16, 17 | 2592 | 40124720 | ||
BX647823.1 | - | 17, 18 | 2966 | |||
BX648157.1 | - | 11, 12 | 1701 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
