siRNA Details
Gene Symbol: FAM65C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290268.1 | NP_001277197.1 | 7 | 851 | ||
NM_080829.3 | NP_543019.2 | 7 | 784 | |||
NR_110890.1 | 7 | 1036 | ||||
XM_005260294.3 | XP_005260351.1 | 8 | 848 | |||
XM_006723713.3 | XP_006723776.1 | 7 | 896 | |||
XM_011528578.2 | XP_011526880.1 | 8 | 876 | |||
XM_011528579.2 | XP_011526881.1 | 8 | 1014 | |||
XM_011528580.2 | XP_011526882.1 | 8 | 1068 | |||
XM_011528581.2 | XP_011526883.1 | 8 | 1052 | |||
XM_011528584.2 | XP_011526886.1 | 7 | 913 | |||
XM_011528586.2 | XP_011526888.1 | 7 | 914 | |||
XM_017027682.1 | XP_016883171.1 | 7 | 913 | |||
GenBank mRNA | AK056792.1 | - | 7 | 755 | ||
AK295781.1 | - | 7 | 727 | |||
AK299337.1 | - | 7 | 700 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
