siRNA Details
Gene Symbol: FAM120A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286722.1 | NP_001273651.1 | 7 | 1454 | ||
NM_001286723.1 | NP_001273652.1 | 7 | 1457 | |||
NM_001286724.1 | NP_001273653.1 | 7 | 1457 | |||
NM_014612.4 | NP_055427.2 | 7 | 1457 | |||
XM_005251842.4 | XP_005251899.1 | 7 | 1409 | |||
XM_011518412.2 | XP_011516714.1 | 7 | 1274 | |||
XM_011518413.2 | XP_011516715.1 | 7 | 1274 | |||
XM_011518414.2 | XP_011516716.1 | 7 | 1271 | |||
XM_011518416.2 | XP_011516718.1 | 7 | 1275 | |||
GenBank mRNA | AF214737.1 | - | 7 | 1262 | ||
AF214738.1 | - | 7 | 1262 | |||
AY266457.1 | - | 1 | 62 | |||
BC007879.2 | - | 6 | 769 | |||
BC098584.1 | - | 7 | 1433 | 30531603 | ||
BC111736.1 | - | 7 | 1255 | 40036337 | ||
D80005.2 | - | 7 | 1218 |
Gene Symbol: C9orf129 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098808.1 | NP_001092278.1 | 3 | 548 | ||
XM_006717117.3 | XP_006717180.1 | 2 | 567 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
