siRNA Details
Gene Symbol: ARID4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206794.1 | NP_001193723.1 | 20 | 3656 | ||
NM_016374.5 | NP_057458.4 | 20 | 3777 | |||
NM_031371.3 | NP_112739.2 | 19 | 3519 | |||
NR_135074.1 | 20 | 3779 | ||||
XM_006711781.2 | XP_006711844.1 | 20 | 3692 | |||
XM_011544212.2 | XP_011542514.1 | 20 | 3616 | |||
XM_017001468.1 | XP_016856957.1 | 19 | 3438 | |||
XM_017001469.1 | XP_016856958.1 | 19 | 3331 | |||
XM_017001470.1 | XP_016856959.1 | 18 | 3179 | |||
XM_017001471.1 | XP_016856960.1 | 18 | 3183 | |||
XM_017001472.1 | XP_016856961.1 | 10 | 2450 | |||
GenBank mRNA | AB210032.1 | - | 20 | 3656 | ||
AF083249.1 | - | 4 | 1640 | |||
AF208045.2 | - | 18 | 3077 | |||
AF214114.2 | - | 19 | 3485 | |||
AF227899.1 | - | 6 | 1962 | |||
AK023144.1 | - | 1 | 799 | |||
AY220790.1 | - | 18 | 3279 | |||
BC130418.1 | - | 20 | 3694 | 40146449 | ||
BX648820.1 | - | 19 | 3471 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
