siRNA Details
Gene Symbol: DNAJC25-GNG10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004125.3 | NP_004116.2 | 1 | 292 | ||
GenBank mRNA | BC015391.1 | - | 1 | 258 | 4425229 | |
BT007068.1 | - | 1 | 236 |
Gene Symbol: DNAJC25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015882.2 | NP_001015882.2 | 1 | 292 | ||
NR_037148.1 | 1 | 292 | ||||
GenBank mRNA | AF447177.1 | - | 1 | 261 | ||
AK095452.1 | - | 1 | 275 | |||
BC048318.1 | - | 1 | 263 | 4456137 | ||
DR004317.1 | - | 1 | 299 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
