siRNA Details
Gene Symbol: DNAJC25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015882.2 | NP_001015882.2 | 3 | 911 | ||
NR_037148.1 | 4 | 1149 | ||||
GenBank mRNA | AF447177.1 | - | 4 | 1118 | ||
AK095452.1 | - | 4 | 1132 | |||
BC048318.1 | - | 3 | 882 | 4456137 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
