siRNA Details
Gene Symbol: DNAJC25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015882.2 | NP_001015882.2 | 4 | 1253 | ||
NR_037148.1 | 5 | 1491 | ||||
GenBank mRNA | AF447177.1 | - | 5 | 1460 | ||
AK095452.1 | - | 5 | 1474 | |||
BC048318.1 | - | 4 | 1224 | 4456137 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
