siRNA Details
Gene Symbol: LZTS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021020.3 | NP_066300.1 | 1 | 410 | ||
XM_005273394.3 | XP_005273451.1 | 2 | 675 | |||
XM_011544383.2 | XP_011542685.1 | 2 | 2613 | |||
XM_011544384.2 | XP_011542686.1 | 2 | 563 | |||
XM_011544385.2 | XP_011542687.1 | 2 | 537 | |||
XM_011544386.2 | XP_011542688.1 | 2 | 762 | |||
GenBank mRNA | AF123654.1 | - | 1 | 299 | ||
AF123655.1 | - | 1 | 299 | |||
AF123656.1 | - | 1 | 299 | |||
AF123657.1 | - | 1 | 299 | |||
AF123658.1 | - | 1 | 299 | |||
AF123659.1 | - | 1 | 410 | |||
BC075006.2 | - | 1 | 348 | 30915401 | ||
BC075007.2 | - | 1 | 348 | 30915614 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
