siRNA Details
Gene Symbol: BORCS8-MEF2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005919.3 | NP_005910.1 | 5 | 855 | ||
NR_027307.1 | 7 | 1033 | ||||
NR_027308.1 | 7 | 1033 | ||||
GenBank mRNA | AK301086.1 | - | 7 | 600 | ||
AK316328.1 | - | 7 | 600 | |||
BC126245.1 | - | 4 | 278 | 8991961 | ||
BC136457.1 | - | 3 | 202 | 9020446 | ||
BC171767.1 | - | 3 | 202 | 9054421 | ||
X63380.1 | - | 7 | 570 | |||
X68502.1 | - | 5 | 377 |
Gene Symbol: MEF2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145785.1 | NP_001139257.1 | 3 | 242 | ||
GenBank mRNA | AK057210.1 | - | 3 | 216 | ||
DA488022.1 | - | 4 | 360 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
