siRNA Details
Gene Symbol: STAG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282716.1 | NP_001269645.1 | 31, 32 | 3757 | ||
NM_001282717.1 | NP_001269646.1 | 31, 32 | 3841 | |||
NM_001282718.1 | NP_001269647.1 | 29, 30 | 3631 | |||
NM_012447.3 | NP_036579.2 | 31, 32 | 3838 | |||
XM_011515742.1 | XP_011514044.1 | 31, 32 | 3808 | |||
XM_017011683.1 | XP_016867172.1 | 31, 32 | 3688 | |||
XM_017011684.1 | XP_016867173.1 | 31, 32 | 3808 | |||
XM_017011685.1 | XP_016867174.1 | 31, 32 | 3921 | |||
XM_017011686.1 | XP_016867175.1 | 31, 32 | 3685 | |||
XM_017011687.1 | XP_016867176.1 | 29, 30 | 3511 | |||
GenBank mRNA | AJ007798.2 | - | 31, 32 | 3758 | ||
AK292394.1 | - | 31, 32 | 3665 | |||
AK302587.1 | - | 28, 29 | 3249 | |||
AK302693.1 | - | 29, 30 | 3454 | |||
AL833816.1 | - | 24, 25 | 3047 | |||
BC140932.1 | - | 31, 32 | 3597 | 9056744 | ||
BC146806.1 | - | 31, 32 | 3663 | 8860345 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
