siRNA Details
Gene Symbol: STON1-GTF2A1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198593.1 | NP_001185522.1 | 5 | 2413 | ||
NM_001198594.1 | NP_001185523.1 | 4 | 2349 | |||
NM_172311.2 | NP_758515.1 | 5 | 2416 | |||
GenBank mRNA | AF026169.1 | - | 5 | 2416 | ||
AK309740.1 | - | 5 | 2413 | |||
BC101772.1 | - | 4 | 2329 | 8069278 | ||
BC101774.1 | - | 4 | 2329 | 8069280 | ||
BC143520.1 | - | 4 | 2329 | 9052029 | ||
BX091338.1 | - | 4 | 483 |
Gene Symbol: GTF2A1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193487.2 | NP_001180416.1 | 2 | 183 | ||
NM_006872.4 | NP_006863.2 | 3 | 285 | |||
GenBank mRNA | AF106857.1 | - | 3 | 205 | ||
AK223350.1 | - | 3 | 209 | |||
AK302220.1 | - | 2 | 166 | |||
BC025991.1 | - | 3 | 234 | 4826221 | ||
BC064585.1 | - | 3 | 189 | |||
BG717289.1 | - | 3 | 260 | |||
HY049561.1 | - | 3 | 288 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
