siRNA Details
Gene Symbol: SPTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024858.2 | NP_001020029.1 | 33 | 6694 | ||
XM_005268023.4 | XP_005268080.1 | 34 | 6761 | |||
XM_017021612.1 | XP_016877101.1 | 35 | 6909 | |||
XM_017021613.1 | XP_016877102.1 | 34 | 6761 | |||
GenBank mRNA | AB209415.1 | - | 34 | 6783 | ||
AF465439.1 | - | 16 | 2740 | |||
AK126627.1 | - | 2 | 4043 | |||
BX248270.1 | - | 3 | 377 | |||
M37884.1 | - | 19 | 3478 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
