siRNA Details
Gene Symbol: SPTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000347.5 | NP_000338.3 | 21 | 4595 | ||
NM_001024858.2 | NP_001020029.1 | 21 | 4595 | |||
XM_005268023.4 | XP_005268080.1 | 22 | 4662 | |||
XM_011537105.2 | XP_011535407.1 | 22 | 4662 | |||
XM_017021612.1 | XP_016877101.1 | 23 | 4810 | |||
XM_017021613.1 | XP_016877102.1 | 22 | 4662 | |||
XM_017021614.1 | XP_016877103.1 | 22 | 4662 | |||
GenBank mRNA | AB209415.1 | - | 22 | 4684 | ||
AF465439.1 | - | 3 | 536 | |||
BC136284.1 | - | 21 | 4574 | 9020271 | ||
BC136285.1 | - | 21 | 4574 | 9020272 | ||
J05500.1 | - | 22 | 4636 | |||
M37884.1 | - | 7 | 1379 | |||
M57948.1 | - | 8 | 1762 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
