siRNA Details
Gene Symbol: GLI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005270.4 | NP_005261.2 | 1 | 119 | ||
XM_006712422.2 | XP_006712485.1 | 1 | 119 | |||
XM_011510974.2 | XP_011509276.1 | 1 | 65 | |||
GenBank mRNA | AB007295.1 | - | 2 | 373 | ||
AB007296.1 | - | 2 | 373 | |||
AB007297.1 | - | 2 | 373 | |||
AB007298.1 | - | 2 | 373 | |||
AK300071.1 | - | 2 | 178 | |||
AY493737.1 | - | 1 | 65 | |||
AY493738.1 | - | 1 | 65 | |||
AY493739.1 | - | 1 | 65 | |||
BC111410.1 | - | 1 | 114 | |||
D14827.1 | - | 2 | 373 | |||
D14828.1 | - | 1 | 218 | |||
DQ004396.1 | - | 1 | 65 | |||
DQ004397.1 | - | 2 | 158 | |||
DQ004398.1 | - | 2 | 149 | |||
DQ086814.1 | - | 1 | 89 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
