siRNA Details
Gene Symbol: GLI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005270.4 | NP_005261.2 | 7 | 1138 | ||
XM_006712422.2 | XP_006712485.1 | 7 | 1138 | |||
XM_011510969.2 | XP_011509271.2 | 7 | 1770 | |||
XM_011510970.2 | XP_011509272.1 | 7 | 2781 | |||
XM_011510971.2 | XP_011509273.1 | 7 | 2031 | |||
XM_011510972.2 | XP_011509274.2 | 7 | 1168 | |||
XM_011510973.2 | XP_011509275.1 | 6 | 919 | |||
XM_011510974.2 | XP_011509276.1 | 6 | 978 | |||
XM_017003818.1 | XP_016859307.1 | 7 | 1770 | |||
GenBank mRNA | AB007295.1 | - | 4 | 695 | ||
AB007296.1 | - | 4 | 695 | |||
AB007297.1 | - | 4 | 695 | |||
AB007298.1 | - | 4 | 695 | |||
BC111410.1 | - | 7 | 1133 | |||
D14827.1 | - | 4 | 695 | |||
D14828.1 | - | 3 | 540 | |||
DQ004396.1 | - | 6 | 978 | |||
DQ086814.1 | - | 7 | 1108 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
