siRNA Details
Gene Symbol: STXBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127396.2 | NP_001120868.1 | 12 | 1021 | ||
NM_001272034.1 | NP_001258963.1 | 12 | 1063 | |||
NM_006949.3 | NP_008880.2 | 12 | 1030 | |||
NR_073560.1 | 12 | 1030 | ||||
XM_011528210.1 | XP_011526512.1 | 12 | 1026 | |||
XM_011528212.2 | XP_011526514.1 | 12 | 1026 | |||
GenBank mRNA | AB002559.1 | - | 12 | 1005 | ||
AK222967.1 | - | 12 | 1007 | |||
AK222977.1 | - | 12 | 1007 | |||
AK302257.1 | - | 12 | 883 | |||
AK303701.1 | - | 12 | 1051 | |||
AK314230.1 | - | 12 | 1009 | |||
BC002869.2 | - | 12 | 995 | 3942156 | ||
BT006915.1 | - | 12 | 972 | |||
U63533.1 | - | 12 | 1005 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
