siRNA Details
Gene Symbol: STXBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127396.2 | NP_001120868.1 | 2, 3 | 143 | ||
NM_001272034.1 | NP_001258963.1 | 2, 3 | 143 | |||
NM_006949.3 | NP_008880.2 | 2, 3 | 143 | |||
NR_073560.1 | 2, 3 | 143 | ||||
XM_011528210.1 | XP_011526512.1 | 2, 3 | 139 | |||
XM_011528212.2 | XP_011526514.1 | 2, 3 | 139 | |||
GenBank mRNA | AB002559.1 | - | 2, 3 | 118 | ||
AK222967.1 | - | 2, 3 | 120 | |||
AK222977.1 | - | 2, 3 | 120 | |||
AK302257.1 | - | 2, 3 | 98 | |||
AK303701.1 | - | 2, 3 | 131 | |||
AK314230.1 | - | 2, 3 | 122 | |||
AU098728.1 | - | 2, 3 | 141 | |||
BC002869.2 | - | 2, 3 | 117 | 3942156 | ||
BQ067493.1 | - | 2, 3 | 144 | |||
BT006915.1 | - | 2, 3 | 94 | |||
DA431480.1 | - | 2, 3 | 139 | |||
U63533.1 | - | 2, 3 | 118 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
