siRNA Details
Gene Symbol: STXBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006949.3 | NP_008880.2 | 4, 5 | 296 | ||
NR_073560.1 | 4, 5 | 296 | ||||
XM_011528210.1 | XP_011526512.1 | 4, 5 | 292 | |||
XM_011528212.2 | XP_011526514.1 | 4, 5 | 292 | |||
GenBank mRNA | AB002559.1 | - | 4, 5 | 271 | ||
AK222967.1 | - | 4, 5 | 273 | |||
AK222977.1 | - | 4, 5 | 273 | |||
AK302257.1 | - | 4, 5 | 251 | |||
AK314230.1 | - | 4, 5 | 275 | |||
BQ067493.1 | - | 4, 5 | 297 | |||
DA431480.1 | - | 4, 5 | 292 | |||
U63533.1 | - | 4, 5 | 271 |

More Information
Other Species Targeted

Gene Ontology Categories:
