siRNA Details
Gene Symbol: TNRC6C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142640.1 | NP_001136112.1 | 16 | 4605 | ||
NM_018996.3 | NP_061869.2 | 16 | 4614 | |||
XM_006721996.3 | XP_006722059.3 | 17 | 10266 | |||
XM_006721997.3 | XP_006722060.3 | 16 | 10116 | |||
XM_006722000.3 | XP_006722063.3 | 17 | 10257 | |||
XM_011525088.2 | XP_011523390.1 | 14 | 4303 | |||
XM_017024906.1 | XP_016880395.1 | 17 | 10257 | |||
XM_017024907.1 | XP_016880396.1 | 17 | 10266 | |||
XM_017024908.1 | XP_016880397.1 | 16 | 10041 | |||
GenBank mRNA | AB046802.2 | - | 13 | 4164 | ||
AK056421.1 | - | 5 | 580 | |||
AL834429.1 | - | 13 | 2136 | |||
BC045631.1 | - | 13 | 1826 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
