siRNA Details
Gene Symbol: XRCC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021141.3 | NP_066964.1 | 5 | 535 | ||
GenBank mRNA | AK096408.1 | - | 2 | 937 | ||
AK290740.1 | - | 5 | 421 | |||
BC019027.2 | - | 5 | 403 | 4555162 | ||
BC095442.1 | - | 5 | 421 | 30329657 | ||
J04977.1 | - | 5 | 428 | |||
M30938.1 | - | 5 | 422 | |||
X57500.1 | - | 2 | 84 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
