siRNA Details
Gene Symbol: XRCC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021141.3 | NP_066964.1 | 11 | 1331 | ||
GenBank mRNA | AK026166.1 | - | 6 | 567 | ||
AK096408.1 | - | 8 | 1733 | |||
AK222603.1 | - | 11 | 1248 | |||
AK290740.1 | - | 11 | 1217 | |||
BC019027.2 | - | 11 | 1199 | 4555162 | ||
BC095442.1 | - | 11 | 1217 | 30329657 | ||
J04977.1 | - | 11 | 1224 | |||
M30938.1 | - | 11 | 1218 | |||
X57500.1 | - | 8 | 880 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
