siRNA Details
Gene Symbol: RIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177663.1 | NP_001171134.1 | 30 | 6205 | ||
NM_001177664.1 | NP_001171135.1 | 30 | 6108 | |||
NM_001177665.1 | NP_001171136.1 | 29 | 6378 | |||
NM_018151.4 | NP_060621.3 | 30 | 6205 | |||
XM_005246665.3 | XP_005246722.1 | 30 | 6085 | |||
XM_017004422.1 | XP_016859911.1 | 30 | 6154 | |||
XM_017004423.1 | XP_016859912.1 | 29 | 5992 | |||
XM_017004424.1 | XP_016859913.1 | 29 | 6022 | |||
XM_017004425.1 | XP_016859914.1 | 28 | 6330 | |||
GenBank mRNA | AK000323.1 | - | 1 | 380 | ||
AL080129.1 | - | 1 | 865 | |||
AY584066.1 | - | 29 | 6044 | |||
AY585745.1 | - | 30 | 6154 | |||
AY727910.1 | - | 30 | 6205 | |||
AY727911.1 | - | 30 | 6108 | |||
AY727912.1 | - | 29 | 6378 | |||
AY727913.1 | - | 30 | 6205 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
