siRNA Details
Gene Symbol: EXD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193360.1 | NP_001180289.1 | 10 | 2951 | ||
NM_001193361.1 | NP_001180290.1 | 9 | 2867 | |||
NM_001193362.1 | NP_001180291.1 | 10 | 3110 | |||
NM_001193363.1 | NP_001180292.1 | 9 | 3026 | |||
NM_018199.3 | NP_060669.1 | 8 | 2487 | |||
NR_034164.1 | 11 | 3041 | ||||
NR_034165.1 | 11 | 3200 | ||||
XM_005267817.3 | XP_005267874.1 | 8 | 2417 | |||
XM_011536908.2 | XP_011535210.1 | 8 | 2505 | |||
XM_017021421.1 | XP_016876910.1 | 9 | 2550 | |||
GenBank mRNA | AK001600.1 | - | 8 | 2415 | ||
AK056025.1 | - | 11 | 2987 | |||
AL834434.1 | - | 8 | 2409 | |||
BX647767.1 | - | 9 | 2817 | |||
CR936728.1 | - | 10 | 2826 | |||
CR936729.1 | - | 9 | 2834 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
