siRNA Details
Gene Symbol: ESF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276380.1 | NP_001263309.1 | 9 | 1891 | ||
NM_016649.3 | NP_057733.2 | 9 | 1902 | |||
XM_017027874.1 | XP_016883363.1 | 9 | 3354 | |||
GenBank mRNA | AF068285.1 | - | 8 | 1259 | ||
AK000375.1 | - | 9 | 1903 | |||
AK074299.1 | - | 8 | 1388 | |||
BC020952.1 | - | 8 | 1521 |
