siRNA Details
Gene Symbol: BCORL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184772.2 | NP_001171701.1 | 7 | 4308 | ||
NM_021946.4 | NP_068765.3 | 7 | 4308 | |||
XM_005262452.4 | XP_005262509.2 | 9 | 4724 | |||
XM_005262453.4 | XP_005262510.2 | 9 | 4492 | |||
XM_005262454.3 | XP_005262511.2 | 8 | 4436 | |||
XM_005262455.4 | XP_005262512.2 | 8 | 4379 | |||
XM_005262456.4 | XP_005262513.2 | 8 | 4334 | |||
XM_006724776.3 | XP_006724839.1 | 9 | 4509 | |||
XM_006724777.3 | XP_006724840.1 | 8 | 4404 | |||
XM_006724779.2 | XP_006724842.1 | 9 | 4595 | |||
XM_017029721.1 | XP_016885210.1 | 9 | 4755 | |||
XM_017029722.1 | XP_016885211.1 | 8 | 4809 | |||
GenBank mRNA | AK074089.1 | - | 7 | 4221 | ||
AK124676.1 | - | 2 | 472 | |||
AL096777.1 | - | 4 | 2180 | |||
BC044652.1 | - | 1 | 94 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
