siRNA Details
Gene Symbol: TAF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015892.1 | NP_001015892.1 | 2 | 399 | ||
NM_003187.4 | NP_003178.1 | 2 | 162 | |||
GenBank mRNA | AI879232.1 | - | 2 | 119 | ||
BC003400.1 | - | 2 | 90 | 3448836 | ||
BC007349.2 | - | 2 | 87 | 2966411 | ||
BC033320.1 | - | 2 | 950 | 4837792 | ||
BG490028.1 | - | 2 | 222 | |||
BG720149.1 | - | 2 | 142 | |||
CX784292.1 | - | 2 | 201 | |||
U21858.1 | - | 2 | 79 |
Gene Symbol: AK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015891.1 | NP_001015891.1 | 2 | 399 | ||
NM_016283.4 | NP_057367.1 | 2 | 162 | |||
GenBank mRNA | AF110777.1 | - | 3 | 411 | ||
AF151895.1 | - | 2 | 102 | |||
AJ878880.1 | - | 2 | 91 | |||
AJ878881.1 | - | 2 | 91 | |||
BC007426.2 | - | 2 | 87 | 2966411 | ||
BX117839.1 | - | 2 | 399 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
