siRNA Details
Gene Symbol: AK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015891.1 | NP_001015891.1 | 2 | 422 | ||
NM_016283.4 | NP_057367.1 | 2 | 185 | |||
GenBank mRNA | AF110777.1 | - | 3 | 434 | ||
AF151895.1 | - | 2 | 125 | |||
AJ878880.1 | - | 2 | 114 | |||
AJ878881.1 | - | 2 | 114 | |||
BC007426.2 | - | 2 | 110 | 2966411 | ||
BX117839.1 | - | 2 | 422 |
Gene Symbol: TAF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015892.1 | NP_001015892.1 | 2 | 422 | ||
NM_003187.4 | NP_003178.1 | 2 | 185 | |||
GenBank mRNA | AI879232.1 | - | 2 | 142 | ||
BC003400.1 | - | 2 | 113 | 3448836 | ||
BC007349.2 | - | 2 | 110 | 2966411 | ||
BC033320.1 | - | 2 | 973 | 4837792 | ||
BG490028.1 | - | 2 | 245 | |||
BG720149.1 | - | 2 | 165 | |||
CX784292.1 | - | 2 | 224 | |||
U21858.1 | - | 2 | 102 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
