siRNA Details
Gene Symbol: SRRT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128852.1 | NP_001122324.1 | 20 | 2970 | ||
NM_001128853.1 | NP_001122325.1 | 20 | 2961 | |||
NM_001128854.1 | NP_001122326.1 | 20 | 2958 | |||
NM_015908.5 | NP_056992.4 | 20 | 2973 | |||
XM_005250405.2 | XP_005250462.1 | 20 | 2830 | |||
XM_005250406.2 | XP_005250463.1 | 20 | 2827 | |||
XM_005250407.2 | XP_005250464.1 | 20 | 2818 | |||
XM_005250408.2 | XP_005250465.1 | 20 | 2815 | |||
XM_017012290.1 | XP_016867779.1 | 21 | 3143 | |||
XM_017012291.1 | XP_016867780.1 | 21 | 3140 | |||
GenBank mRNA | AF082871.1 | - | 10 | 1369 | ||
AF112994.1 | - | 7 | 1158 | |||
AF248955.1 | - | 20 | 2924 | |||
AK296131.1 | - | 14 | 1939 | |||
AK302820.1 | - | 20 | 2862 | |||
AL096723.1 | - | 20 | 2924 | |||
BC000082.2 | - | 11 | 1494 | |||
BC069249.1 | - | 18 | 2514 | |||
BC109117.1 | - | 20 | 2852 | 40035609 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
