siRNA Details
Gene Symbol: CFAP61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167816.1 | NP_001161288.1 | 4, 5 | 455 | ||
NM_015585.3 | NP_056400.3 | 5, 6 | 561 | |||
XM_005260688.3 | XP_005260745.1 | 5, 6 | 524 | |||
XM_005260689.2 | XP_005260746.1 | 5, 6 | 524 | |||
XM_005260690.3 | XP_005260747.1 | 5, 6 | 524 | |||
XM_005260691.3 | XP_005260748.1 | 5, 6 | 524 | |||
XM_005260692.3 | XP_005260749.1 | 5, 6 | 524 | |||
XM_005260693.3 | XP_005260750.1 | 5, 6 | 524 | |||
XM_005260697.1 | XP_005260754.1 | 5, 6 | 524 | |||
XM_011529211.1 | XP_011527513.1 | 4, 5 | 382 | |||
XM_011529212.2 | XP_011527514.1 | 5, 6 | 524 | |||
XM_011529213.1 | XP_011527515.1 | 5, 6 | 524 | |||
XM_017027786.1 | XP_016883275.1 | 5, 6 | 524 | |||
XM_017027787.1 | XP_016883276.1 | 5, 6 | 524 | |||
XM_017027788.1 | XP_016883277.1 | 5, 6 | 524 | |||
XM_017027789.1 | XP_016883278.1 | 5, 6 | 524 | |||
XM_017027790.1 | XP_016883279.1 | 5, 6 | 524 | |||
XM_017027795.1 | XP_016883284.1 | 5, 6 | 524 | |||
GenBank mRNA | AK304174.1 | - | 5, 6 | 524 | ||
AK307817.1 | - | 4, 5 | 382 | |||
AL117439.1 | - | 4, 5 | 393 | |||
BC028708.2 | - | 5, 6 | 523 | 4825941 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
