siRNA Details
Gene Symbol: CMTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052999.3 | NP_443725.3 | 4 | 964 | ||
NM_181268.2 | NP_851785.2 | 4 | 948 | |||
NM_181269.2 | NP_851786.1 | 4 | 669 | |||
NM_181270.2 | NP_851787.1 | 4 | 653 | |||
NM_181271.2 | NP_851788.1 | 3 | 528 | |||
NM_181272.2 | NP_851789.1 | 3 | 510 | |||
NM_181283.2 | NP_851800.1 | 3 | 494 | |||
NM_181296.2 | NP_851813.1 | 2 | 395 | |||
GenBank mRNA | AF278576.1 | - | 4 | 568 | ||
AF278577.1 | - | 4 | 584 | |||
AY174118.1 | - | 4 | 673 | |||
AY174119.1 | - | 4 | 657 | |||
AY174120.1 | - | 3 | 532 | |||
AY174121.1 | - | 3 | 514 | |||
AY174122.1 | - | 3 | 498 | |||
AY174123.1 | - | 3 | 669 | |||
AY174124.1 | - | 3 | 653 | |||
AY174125.1 | - | 5 | 807 | |||
AY174126.1 | - | 5 | 791 | |||
AY174127.1 | - | 2 | 361 | |||
AY174128.1 | - | 4 | 637 | |||
AY174129.1 | - | 5 | 660 | |||
AY174130.1 | - | 5 | 832 | |||
AY174131.1 | - | 3 | 489 | |||
AY174132.1 | - | 3 | 473 | |||
AY174133.1 | - | 2 | 399 | |||
AY174134.1 | - | 4 | 719 | |||
AY174135.1 | - | 5 | 897 | |||
AY174136.1 | - | 5 | 881 | |||
AY174137.1 | - | 4 | 699 | |||
AY174138.1 | - | 4 | 683 | |||
BC057852.1 | - | 4 | 964 | 5273108 | ||
DQ309766.1 | - | 5 | 782 | |||
DQ355985.1 | - | 3 | 520 |
Gene Symbol: CKLF-CMTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202509.1 | NP_001189438.1 | 3 | 537 | ||
NM_001204098.1 | NP_001191027.1 | 4 | 617 | |||
NM_001204099.1 | NP_001191028.1 | 4 | 696 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
