siRNA Details
Gene Symbol: CMTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052999.3 | NP_443725.3 | 4 | 1041 | ||
NM_181268.2 | NP_851785.2 | 4 | 1025 | |||
NM_181269.2 | NP_851786.1 | 4 | 746 | |||
NM_181270.2 | NP_851787.1 | 4 | 730 | |||
NM_181271.2 | NP_851788.1 | 3 | 605 | |||
NM_181272.2 | NP_851789.1 | 3 | 587 | |||
NM_181283.2 | NP_851800.1 | 3 | 571 | |||
NM_181296.2 | NP_851813.1 | 2 | 472 | |||
GenBank mRNA | AF278576.1 | - | 4 | 645 | ||
AF278577.1 | - | 4 | 661 | |||
AY174118.1 | - | 4 | 750 | |||
AY174119.1 | - | 4 | 734 | |||
AY174120.1 | - | 3 | 609 | |||
AY174121.1 | - | 3 | 591 | |||
AY174122.1 | - | 3 | 575 | |||
AY174123.1 | - | 3 | 746 | |||
AY174124.1 | - | 3 | 730 | |||
AY174125.1 | - | 5 | 884 | |||
AY174126.1 | - | 5 | 868 | |||
AY174127.1 | - | 2 | 438 | |||
AY174128.1 | - | 4 | 714 | |||
AY174129.1 | - | 5 | 737 | |||
AY174130.1 | - | 5 | 909 | |||
AY174131.1 | - | 3 | 566 | |||
AY174132.1 | - | 3 | 550 | |||
AY174133.1 | - | 2 | 476 | |||
AY174134.1 | - | 4 | 796 | |||
AY174135.1 | - | 5 | 974 | |||
AY174136.1 | - | 5 | 958 | |||
AY174137.1 | - | 4 | 776 | |||
AY174138.1 | - | 4 | 760 | |||
BC057852.1 | - | 4 | 1041 | 5273108 | ||
DQ309766.1 | - | 5 | 859 | |||
DQ355985.1 | - | 3 | 597 |
Gene Symbol: CKLF-CMTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202509.1 | NP_001189438.1 | 3 | 614 | ||
NM_001204098.1 | NP_001191027.1 | 4 | 694 | |||
NM_001204099.1 | NP_001191028.1 | 4 | 773 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
