siRNA Details
Gene Symbol: LTBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042544.1 | NP_001036009.1 | 26, 27 | 3691 | ||
NM_001042545.1 | NP_001036010.1 | 23, 24 | 3507 | |||
NM_003573.2 | NP_003564.2 | 26, 27 | 3580 | |||
XM_011527376.2 | XP_011525678.1 | 25, 26 | 3834 | |||
XM_011527377.2 | XP_011525679.1 | 26, 27 | 3739 | |||
XM_011527379.1 | XP_011525681.1 | 22, 23 | 3642 | |||
XM_011527380.2 | XP_011525682.1 | 25, 26 | 3613 | |||
XM_011527381.2 | XP_011525683.1 | 25, 26 | 3613 | |||
XM_011527382.2 | XP_011525684.1 | 25, 26 | 3496 | |||
XM_011527387.1 | XP_011525689.1 | 26, 27 | 3839 | |||
XM_017027352.1 | XP_016882841.1 | 25, 26 | 3607 | |||
XM_017027353.1 | XP_016882842.1 | 24, 25 | 3481 | |||
GenBank mRNA | AF051344.2 | - | 23, 24 | 3507 | ||
AF051345.2 | - | 26, 27 | 3691 | |||
AF054502.1 | - | 8, 9 | 1104 | |||
AK074499.1 | - | 4, 5 | 995 | |||
AK128191.1 | - | 12, 13 | 1689 | |||
Y13622.1 | - | 25, 26 | 3580 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
