siRNA Details
Gene Symbol: LTBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042544.1 | NP_001036009.1 | 19 | 2552 | ||
NM_001042545.1 | NP_001036010.1 | 16 | 2368 | |||
NM_003573.2 | NP_003564.2 | 19 | 2441 | |||
XM_011527376.2 | XP_011525678.1 | 18 | 2695 | |||
XM_011527377.2 | XP_011525679.1 | 19 | 2600 | |||
XM_011527378.2 | XP_011525680.1 | 19 | 2600 | |||
XM_011527379.1 | XP_011525681.1 | 15 | 2503 | |||
XM_011527380.2 | XP_011525682.1 | 18 | 2474 | |||
XM_011527381.2 | XP_011525683.1 | 19 | 2600 | |||
XM_011527382.2 | XP_011525684.1 | 18 | 2357 | |||
XM_011527383.2 | XP_011525685.1 | 19 | 2600 | |||
XM_011527384.2 | XP_011525686.1 | 19 | 2600 | |||
XM_011527385.2 | XP_011525687.1 | 19 | 2600 | |||
XM_011527386.2 | XP_011525688.1 | 19 | 2600 | |||
XM_011527387.1 | XP_011525689.1 | 19 | 2700 | |||
XM_017027352.1 | XP_016882841.1 | 19 | 2600 | |||
XM_017027353.1 | XP_016882842.1 | 19 | 2600 | |||
XM_017027354.1 | XP_016882843.1 | 18 | 2474 | |||
GenBank mRNA | AF051344.2 | - | 16 | 2368 | ||
AF051345.2 | - | 19 | 2552 | |||
AF054502.1 | - | 3 | 223 | |||
AK128191.1 | - | 5 | 550 | |||
AK303613.1 | - | 4 | 784 | |||
AK309484.1 | - | 5 | 749 | |||
Y13622.1 | - | 18 | 2441 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
