siRNA Details
Gene Symbol: LTBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042544.1 | NP_001036009.1 | 31 | 4289 | ||
NM_001042545.1 | NP_001036010.1 | 28 | 4105 | |||
NM_003573.2 | NP_003564.2 | 31 | 4178 | |||
XM_011527376.2 | XP_011525678.1 | 30 | 4432 | |||
XM_011527377.2 | XP_011525679.1 | 31 | 4337 | |||
XM_011527378.2 | XP_011525680.1 | 30 | 4265 | |||
XM_011527379.1 | XP_011525681.1 | 27 | 4240 | |||
XM_011527380.2 | XP_011525682.1 | 30 | 4211 | |||
XM_011527381.2 | XP_011525683.1 | 30 | 4211 | |||
XM_011527382.2 | XP_011525684.1 | 30 | 4094 | |||
XM_011527383.2 | XP_011525685.1 | 29 | 3998 | |||
XM_011527384.2 | XP_011525686.1 | 28 | 3872 | |||
XM_011527385.2 | XP_011525687.1 | 28 | 3866 | |||
XM_011527386.2 | XP_011525688.1 | 27 | 3740 | |||
XM_011527387.1 | XP_011525689.1 | 31 | 4437 | |||
XM_017027352.1 | XP_016882841.1 | 30 | 4205 | |||
XM_017027353.1 | XP_016882842.1 | 29 | 4079 | |||
XM_017027354.1 | XP_016882843.1 | 27 | 3746 | |||
GenBank mRNA | AF051344.2 | - | 28 | 4105 | ||
AF051345.2 | - | 31 | 4289 | |||
AF054502.1 | - | 13 | 1702 | |||
AK074499.1 | - | 9 | 1593 | |||
AK128191.1 | - | 17 | 2287 | |||
Y13622.1 | - | 30 | 4178 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
