siRNA Details
Gene Symbol: CD209 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144893.1 | NP_001138365.1 | 2 | 274 | ||
NM_001144894.1 | NP_001138366.1 | 2 | 274 | |||
NM_001144895.1 | NP_001138367.1 | 4 | 406 | |||
NM_001144896.1 | NP_001138368.1 | 3 | 334 | |||
NM_001144897.1 | NP_001138369.1 | 4 | 406 | |||
NM_021155.3 | NP_066978.1 | 4 | 406 | |||
NR_026692.1 | 3 | 506 | ||||
GenBank mRNA | AF290886.1 | - | 4 | 348 | ||
AK293089.1 | - | 4 | 362 | |||
AK308595.1 | - | 1 | 87 | |||
AK313585.1 | - | 4 | 407 | |||
AY042221.1 | - | 4 | 339 | |||
AY042222.1 | - | 4 | 339 | |||
AY042223.1 | - | 4 | 339 | |||
AY042225.1 | - | 3 | 267 | |||
AY042226.1 | - | 2 | 207 | |||
AY042227.1 | - | 2 | 207 | |||
AY042229.1 | - | 3 | 439 | |||
AY042230.1 | - | 2 | 367 | |||
AY042231.1 | - | 2 | 367 | |||
AY042232.1 | - | 2 | 367 | |||
AY042233.1 | - | 2 | 367 | |||
BC110615.2 | - | 2 | 374 | 40032459 | ||
M98457.1 | - | 4 | 380 |
Gene Symbol: CLEC4M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC110614.2 | - | 2 | 348 | 40032453 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
