siRNA Details
Gene Symbol: CD209 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144893.1 | NP_001138365.1 | 4 | 593 | ||
NM_001144894.1 | NP_001138366.1 | 4 | 869 | |||
NM_001144895.1 | NP_001138367.1 | 6 | 725 | |||
NM_001144896.1 | NP_001138368.1 | 5 | 929 | |||
NM_001144897.1 | NP_001138369.1 | 6 | 983 | |||
NM_001144899.1 | NP_001138371.1 | 6 | 518 | |||
NM_021155.3 | NP_066978.1 | 6 | 1001 | |||
NR_026692.1 | 5 | 1101 | ||||
XM_005272472.4 | XP_005272529.1 | 6 | 618 | |||
GenBank mRNA | AF290886.1 | - | 6 | 943 | ||
AK293089.1 | - | 6 | 957 | |||
AK304190.1 | - | 6 | 504 | |||
AK308595.1 | - | 3 | 544 | |||
AK313585.1 | - | 6 | 1002 | |||
AY042221.1 | - | 6 | 934 | |||
AY042222.1 | - | 6 | 916 | |||
AY042223.1 | - | 6 | 658 | |||
AY042224.1 | - | 4, 5 | 226 | |||
AY042225.1 | - | 5 | 862 | |||
AY042226.1 | - | 4 | 802 | |||
AY042227.1 | - | 5 | 526 | |||
AY042229.1 | - | 5 | 1034 | |||
AY042230.1 | - | 4 | 962 | |||
AY042231.1 | - | 4 | 824 | |||
BC110615.2 | - | 4 | 969 | 40032459 | ||
M98457.1 | - | 6 | 975 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
