siRNA Details
Gene Symbol: WAC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016628.4 | NP_057712.2 | 8 | 1045 | ||
NM_100264.2 | NP_567822.1 | 8 | 1336 | |||
NM_100486.3 | NP_567823.1 | 7 | 736 | |||
XM_011519491.2 | XP_011517793.1 | 8 | 1646 | |||
XM_017016315.1 | XP_016871804.1 | 8 | 1156 | |||
XM_017016316.1 | XP_016871805.1 | 8 | 1646 | |||
XM_017016317.1 | XP_016871806.1 | 7 | 847 | |||
XM_017016318.1 | XP_016871807.1 | 7 | 847 | |||
GenBank mRNA | AB058747.1 | - | 6 | 755 | ||
AF116666.2 | - | 9 | 1110 | |||
AK055852.1 | - | 8 | 1153 | |||
AK223600.1 | - | 8 | 1043 | |||
AK225846.1 | - | 8 | 1149 | |||
AK290174.1 | - | 8 | 1050 | |||
AK297460.1 | - | 8 | 958 | |||
AL713727.1 | - | 6 | 1175 | |||
BC004258.1 | - | 8 | 1036 | 3347345 | ||
BC010356.1 | - | 3 | 187 |

More Information
Other Species Targeted

Gene Ontology Categories:
