siRNA Details
Gene Symbol: LTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002341.1 | NP_002332.1 | 4 | 357 | ||
NM_009588.1 | NP_033666.1 | 3 | 311 | |||
GenBank mRNA | BC069330.1 | - | 4 | 357 | 7262211 | |
L11015.1 | - | 4 | 357 | |||
U79029.1 | - | 3 | 175 | |||
U89922.1 | - | 3 | 311 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
