siRNA Details
Gene Symbol: CECR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290046.1 | NP_001276975.1 | 19 | 5282 | ||
NM_001290047.1 | NP_001276976.1 | 19 | 5754 | |||
XM_006724079.3 | XP_006724142.1 | 7 | 4197 | |||
XM_011546128.2 | XP_011544430.1 | 18 | 5358 | |||
XM_011546129.2 | XP_011544431.1 | 19 | 5358 | |||
XM_011546132.2 | XP_011544434.1 | 19 | 5277 | |||
XM_017028785.1 | XP_016884274.1 | 19 | 5439 | |||
GenBank mRNA | AF336133.1 | - | 19 | 5814 | ||
AK091754.1 | - | 4 | 1792 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
