siRNA Details
Gene Symbol: GTPBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004286.4 | NP_004277.2 | 12 | 2853 | ||
GenBank mRNA | AF370385.1 | - | 1 | 426 | ||
AK095950.1 | - | 1 | 414 | |||
AK096647.1 | - | 2 | 2191 | |||
BC014075.1 | - | 12 | 2693 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
