siRNA Details
Gene Symbol: HOMEZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020834.2 | NP_065885.2 | 2 | 549 | ||
GenBank mRNA | AB037864.1 | - | 2 | 475 | ||
AF463523.1 | - | 2 | 473 | |||
AK296927.1 | - | 2 | 509 | |||
AK301849.1 | - | 2 | 395 | |||
AK302788.1 | - | 2 | 409 | |||
BC130392.1 | - | 1 | 374 | 40146423 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
