siRNA Details
Gene Symbol: HOMEZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020834.2 | NP_065885.2 | 2 | 398 | ||
GenBank mRNA | AB037864.1 | - | 2 | 324 | ||
AF463523.1 | - | 2 | 322 | |||
AK296927.1 | - | 2 | 358 | |||
AK301849.1 | - | 2 | 244 | |||
AK302788.1 | - | 2 | 258 | |||
BC130392.1 | - | 1 | 223 | 40146423 | ||
DC398645.1 | - | 2 | 415 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
