siRNA Details
Gene Symbol: MDP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199821.1 | NP_001186750.1 | 5 | 503 | ||
NM_001199822.1 | NP_001186751.1 | 6 | 691 | |||
NM_138476.3 | NP_612485.2 | 6 | 643 | |||
GenBank mRNA | AK092821.1 | - | 4 | 814 | ||
BC009495.2 | - | 4 | 711 | |||
BC046912.1 | - | 5 | 498 | 5287227 | ||
BC051382.1 | - | 6 | 608 | 6268667 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
