siRNA Details
Gene Symbol: OAS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002535.2 | NP_002526.2 | 3 | 739 | ||
NM_016817.2 | NP_058197.2 | 3 | 739 | |||
GenBank mRNA | AK292796.1 | - | 3 | 628 | ||
AK292906.1 | - | 3 | 656 | |||
BC049215.1 | - | 3 | 640 | 5517804 | ||
BQ213828.1 | - | 2 | 266 | |||
M87284.1 | - | 3 | 618 | |||
M87434.1 | - | 3 | 618 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
