siRNA Details
Gene Symbol: MDP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199821.1 | NP_001186750.1 | 1 | 48 | ||
NM_001199822.1 | NP_001186751.1 | 1 | 48 | |||
NM_138476.3 | NP_612485.2 | 1 | 48 | |||
GenBank mRNA | BC046912.1 | - | 1 | 43 | 5287227 | |
BC051382.1 | - | 1 | 13 | 6268667 | ||
DA287183.1 | - | 1 | 48 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
