siRNA Details
Gene Symbol: MDP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199822.1 | NP_001186751.1 | 4 | 411 | ||
NM_138476.3 | NP_612485.2 | 4 | 411 | |||
GenBank mRNA | AK092821.1 | - | 4 | 328 | ||
AK312038.1 | - | 4 | 348 | |||
BC009495.2 | - | 2 | 479 | |||
BC051382.1 | - | 4 | 376 | 6268667 | ||
CB046258.1 | - | 4 | 368 | |||
DA287183.1 | - | 3 | 485 |
Gene Symbol: NEDD8-MDP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CB070148.1 | - | 6 | 456 | ||
DN993851.1 | - | 5 | 437 | |||
FY211956.1 | - | 6 | 426 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
