siRNA Details
Gene Symbol: NINL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318226.1 | NP_001305155.1 | 19 | 2480 | ||
NM_025176.5 | NP_079452.3 | 20 | 3527 | |||
XM_011529186.2 | XP_011527488.1 | 21 | 4366 | |||
XM_011529187.2 | XP_011527489.1 | 21 | 4351 | |||
XM_011529188.1 | XP_011527490.1 | 21 | 3743 | |||
XM_011529189.1 | XP_011527491.1 | 21 | 3751 | |||
XM_011529190.1 | XP_011527492.1 | 19 | 3465 | |||
XM_011529191.1 | XP_011527493.1 | 17 | 3258 | |||
XM_011529192.1 | XP_011527494.1 | 14 | 4178 | |||
XM_011529193.1 | XP_011527495.1 | 18 | 2418 | |||
XM_011529194.2 | XP_011527496.1 | 12 | 3447 | |||
XM_011529195.2 | XP_011527497.1 | 11 | 2272 | |||
XM_011529197.2 | XP_011527499.1 | 13 | 2641 | |||
XM_011529198.1 | XP_011527500.1 | 7 | 1813 | |||
XM_017027732.1 | XP_016883221.1 | 22 | 5685 | |||
XM_017027733.1 | XP_016883222.1 | 22 | 3797 | |||
XM_017027734.1 | XP_016883223.1 | 14 | 2631 | |||
XM_017027735.1 | XP_016883224.1 | 11 | 2277 | |||
GenBank mRNA | AB023197.1 | - | 20 | 3513 | ||
BC036380.2 | - | 19 | 3459 | 5268454 | ||
EU718622.1 | - | 19 | 2466 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
