siRNA Details
Gene Symbol: NINL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318226.1 | NP_001305155.1 | 18, 19 | 2457 | ||
NM_025176.5 | NP_079452.3 | 19, 20 | 3504 | |||
XM_011529186.2 | XP_011527488.1 | 20, 21 | 4343 | |||
XM_011529187.2 | XP_011527489.1 | 20, 21 | 4328 | |||
XM_011529188.1 | XP_011527490.1 | 20, 21 | 3720 | |||
XM_011529189.1 | XP_011527491.1 | 20, 21 | 3728 | |||
XM_011529190.1 | XP_011527492.1 | 18, 19 | 3442 | |||
XM_011529191.1 | XP_011527493.1 | 16, 17 | 3235 | |||
XM_011529192.1 | XP_011527494.1 | 13, 14 | 4155 | |||
XM_011529193.1 | XP_011527495.1 | 17, 18 | 2395 | |||
XM_011529194.2 | XP_011527496.1 | 11, 12 | 3424 | |||
XM_011529195.2 | XP_011527497.1 | 10, 11 | 2249 | |||
XM_011529197.2 | XP_011527499.1 | 12, 13 | 2618 | |||
XM_011529198.1 | XP_011527500.1 | 6, 7 | 1790 | |||
XM_017027732.1 | XP_016883221.1 | 21, 22 | 5662 | |||
XM_017027733.1 | XP_016883222.1 | 21, 22 | 3774 | |||
XM_017027734.1 | XP_016883223.1 | 13, 14 | 2608 | |||
XM_017027735.1 | XP_016883224.1 | 10, 11 | 2254 | |||
GenBank mRNA | AB023197.1 | - | 19, 20 | 3490 | ||
BC036380.2 | - | 18, 19 | 3436 | 5268454 | ||
EU718622.1 | - | 18, 19 | 2443 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
