siRNA Details
Gene Symbol: NINL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318226.1 | NP_001305155.1 | 12 | 1623 | ||
NM_025176.5 | NP_079452.3 | 12 | 1623 | |||
XM_011529186.2 | XP_011527488.1 | 12 | 2261 | |||
XM_011529187.2 | XP_011527489.1 | 12 | 2246 | |||
XM_011529188.1 | XP_011527490.1 | 12 | 1638 | |||
XM_011529189.1 | XP_011527491.1 | 12 | 1646 | |||
XM_011529190.1 | XP_011527492.1 | 11 | 1561 | |||
XM_011529191.1 | XP_011527493.1 | 8 | 1153 | |||
XM_011529192.1 | XP_011527494.1 | 5 | 2073 | |||
XM_011529193.1 | XP_011527495.1 | 11 | 1561 | |||
XM_011529194.2 | XP_011527496.1 | 3 | 1342 | |||
XM_011529195.2 | XP_011527497.1 | 2 | 167 | |||
XM_011529197.2 | XP_011527499.1 | 4 | 536 | |||
XM_017027732.1 | XP_016883221.1 | 13 | 3580 | |||
XM_017027733.1 | XP_016883222.1 | 13 | 1692 | |||
XM_017027734.1 | XP_016883223.1 | 6 | 727 | |||
XM_017027735.1 | XP_016883224.1 | 2 | 172 | |||
GenBank mRNA | AB023197.1 | - | 12 | 1609 | ||
AK093569.1 | - | 2 | 1006 | |||
AK309298.1 | - | 12 | 1572 | |||
BC036380.2 | - | 11 | 1561 | 5268454 | ||
EU718622.1 | - | 12 | 1609 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
