siRNA Details
Gene Symbol: SIRPB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011529130.2 | XP_011527432.1 | 6 | 1705 | ||
GenBank mRNA | AK130124.1 | - | 4 | 965 | ||
AK289866.1 | - | 6 | 1620 | |||
BM285386.1 | - | 3 | 691 | |||
BX537992.1 | - | 6 | 1720 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
